Ultra-Adhesive Tape
Catalog # SAMP8224A-95
Supplier: Sampling Systems
Specifications
- Largeur:95 mm
- Couleur:blanc
- Désignation:Ultra-adhesive tape
- Longueur:50 m
- Cdt:1 Roll
Specifications
About this item
Ultra-adhesive tape is an innovative solution for many applications.
- Can be easily written on
- Available from stock
- Fast delivery
Ultra-adhesive tape offers a wide variety of uses, including applications in warehouse, production facilities, and laboratories. It is ideal for sealing sacks and bags after sampling and dispensing. The tape is also foil-backed to give extra strength to the label.