EpiCult™ Plus Medium
Product Details & Documents
For culture of human, mouse, and rat epithelial stem cells.
EpiCult™ Plus Medium is a serum- and BPE-free culture medium for the expansion of epithelial stem cells from mammary, prostate, and other stratified and pseudostratified epithelial tissues from human, mouse, and rat. Epithelial cells can be expanded in EpiCult™ Plus Medium for multiple passages and for <30 population doublings; for some human mammary cells, addition of fetal bovine serum may be required. EpiCult™ Plus Medium is not recommended for human keratinocytes or primary human airway cells; for culturing primary human airway cells, we recommend PneumaCult™-Ex Plus Medium.
Note: Hydrocortisone stock solution is required for preparation of complete EpiCult™ Plus Medium.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spécifications
Spécifications
- Cdt:500 mL
- Désignation:EpiCult™ Plus medium