Alpha MEM without Nucleosides
Alpha MEM without nucleosides, 500 mL
N° de catalogue STMC36453
Marketsource
Fournisseur:   Stemcell Technologies
Alpha MEM without Nucleosides
N° de catalogue STMC36453
Fournisseur:  Stemcell Technologies
Marketsource
Numéro CAS:  
Alpha MEM without Nucleosides
Alpha MEM without Nucleosides
Alpha MEM without nucleosides, 500 mL
Numéro CAS:  
Prix
63,00 €
Connexion Pour la disponibilité
1 * 500 mL
Product Details & Documents
Minimum Essential Medium Eagle - alpha modification (Alpha MEM) without nucleosides.
Minimum Essential Medium Eagle - Alpha Modification (Alpha MEM) without Nucleosides is recommended for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition. MEM has typically been used for the cultivation of cells grown in monolayers; however, additional supplementation to Alpha MEM has expanded the applications for this medium.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spécifications
Spécifications
  • Cdt:
    500 mL
  • Désignation:
    Alpha MEM without nucleosides
Spécifications