ContactUsLinkComponent
Fetal Bovine Serum, AUS Origin
Fetal Bovine Serum, AUS Origin
  STMC100-0180
 :  
undefined
Fetal Bovine Serum, AUS Origin
  STMC100-0180
 :  
  • Cdt:
    500 mL
  • Désignation:
    Fetal bovine serum, AUS origin, 500 ml
Fetal bovine serum for cell culture and research applications.
STEMCELL fetal bovine serum (FBS) can be used in a broad range of applications including as a serum supplement for culture media or as a research assay reagent. Stemcell FBS is sterile and has low endotoxin and hemoglobin levels.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').