ContactUsLinkComponent
NaïveCult™ Expansion Medium
NaïveCult™ Expansion Medium
N° de catalogue STMC05590
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
undefined
NaïveCult™ Expansion Medium
N° de catalogue STMC05590
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    NaïveCult™ Expansion medium
Spécifications
À propos de cet article
Defined cell culture medium for expansion of reset naïve human embryonic stem (ES) and induced pluripotent stem (iPS) cells.
Robustly expand reset naïve human embryonic stem (ES) and induced pluripotent stem (iPS) cells using a defined cell culture medium. Reset naïve human pluripotent stem cells (hPSCs) cultured in this medium demonstrate uniformly domed, phase-bright colonies with refractive edges and maintain gene expression profiles associated with reset naïve hPSCs. Reset naïve hPSCs cultured in this medium are capable of tri-lineage differentiation into all three somatic lineages following re-priming.
Based on the t2iL + Gö formulation by Austin Smith and developed under license from the Cambridge Stem Cell Institute at the University of Cambridge, UK, this medium is compatible with reset naïve human ES and iPS cells derived using NaïveCult™ Induction Kit, transgene-induced reset human pluripotent stem cells (hPSCs), or embryo-derived reset hPSCs.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').