STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimised)
STEMdiff™ Definitive endoderm kit (TeSR™-E8™ optimised), 1 KIT
N° de catalogue STMC05115
Marketsource
Fournisseur:   Stemcell Technologies
STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimised)
N° de catalogue STMC05115
Fournisseur:  Stemcell Technologies
Marketsource
Numéro CAS:  
STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimised)
STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimised)
STEMdiff™ Definitive endoderm kit (TeSR™-E8™ optimised), 1 KIT
Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Defined animal component-free medium for the differentiation of human ES and iPS cells maintained in TeSR™-E8™ to definitive endoderm.
STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimised) is a complete, serum- and animal component-free medium and supplement kit that supports highly efficient differentiation of human ES cells and iPS cells that have been maintained in TeSR™-E8™ to definitive endoderm. Cells differentiated using the STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimised) express high levels of endoderm markers, including CD184 (CXCR4) and SOX17. When used in combination with Vitronectin XF™, the STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimised) provides a completely xeno-free workflow for the generation of multipotent definitive endoderm. Optimised for differentiation of cells maintained in TeSR™-E8™ . For differentiation of cells maintained in mTeSR™1, please see the STEMdiff™ Definitive Endoderm Kit.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    STEMdiff™ Definitive endoderm kit (TeSR™-E8™ optimised)
Spécifications