ContactUsLinkComponent
mTeSR™ Plus Medium
mTeSR™ Plus Medium
Catalog # STMC100-1130
CAS Number:  
undefined
mTeSR™ Plus Medium
Catalog # STMC100-1130
CAS Number:  
Specifications
  • Cdt:
    1 L
  • Désignation:
    mTeSR™ Plus
Specifications
About this item
cGMP, stabilised feeder-free maintenance medium for human ES and iPS cells.
Enjoy weekend-free schedules and enhanced growth characteristics while maintaining cell quality with this stabilised feeder-free maintenance medium for human pluripotent stem cells (hPSCs). Manufactured under relevant cGMPs, mTeSR™ Plus ensures the highest quality and consistency for fundamental research as well as for cell therapy and investigational new drug research applications. It is based on mTeSR™1, the most widely published feeder-free cell culture medium for hPSCs. With stabilised critical medium components, including FGF2, and enhanced pH buffering, use mTeSR™ Plus to maintain cell-quality attributes and increase cell expansion rates with either daily or restricted feeding. Each lot of mTeSR™ Plus 5X Supplement is used to prepare complete mTeSR™ Plus medium and then performance-tested in a culture assay using human pluripotent stem cells (hPSCs). mTeSR™ Plus is compatible with a variety of culture matrices, including Corning® Matrigel® hESC-qualified matrix and Vitronectin XF™.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').