STEMdiff™ Neural Crest Differentiation Kit
Catalog # STMC08610
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:STEMdiff™ neural crest differentiation kit
Specifications
About this item
Cell culture kit for establishment of hPSC-derived neural crest cells.
STEMdiff™ neural crest differentiation kit creates a serum-free medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to neural crest cells. These neural crest cells, which are characterized by neural crest markers such as SOX10 and CD271, can be differentiated to several downstream derivatives including chondrocytes, osteoblasts, and peripheral neurons. This medium is compatible with human ES and iPS cells maintained in either mTeSR™1, mTeSR™ plus or TeSR™-E8™.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').