TeSR™ E7™ Medium for Reprogramming (2-Component)
STMC05914
:
- Cdt:500 mL
- Désignation:TeSR™-E7™ medium for Reprogramming (2-component)
Feeder-free and animal component-free reprogramming medium for human iPS cell induction.
TeSR™-E7™ (2-component) is an animal component-free and defined reprogramming culture medium optimised for the generation of human iPS cells without the use of feeders. It is based on the E7 formulation.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').