- Cdt:450 mL
- Désignation:NeuroCult™ basal medium (Mouse and Rat)
Basal medium for culture of mouse and rat neural stem and progenitor cells.
NeuroCult™ basal medium (Mouse and Rat) is a standardised serum-free basal medium designed to be supplemented with NeuroCult™ proliferation supplement (Mouse and Rat) and appropriate cytokines for the in vitro culture and expansion of mouse and rat neural stem and progenitor cells. NeuroCult™ basal medium (Mouse and Rat) can also be supplemented with NeuroCult™ differentiation supplement (Mouse and Rat) for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes. NeuroCult™ basal medium (Mouse and Rat) is a component of NeuroCult™ proliferation kit (Mouse and Rat) and NeuroCult™ differentiation kit (Mouse and Rat).
NOTE: When preparing complete NeuroCult™ proliferation medium, addition of human recombinant EGF is required. When culturing cells obtained from adult mouse or rat, human recombinant bFGF and heparin solution are also required.