PolTech Ultra DES Spray
Catalog # BSNC2600041
Supplier: Basan
New Product
Specifications
- Capacité:6×750 ml
- Désignation:PolTech Ultra DES spray
- Cdt:6
- Valeur pH:10,5 ±1
Specifications
About this item
Ready-to-use disinfectant cleaner (bactericidal, yeasticidal and virucidal against enveloped viruses) that allows the cleaning and disinfection of all washable surfaces (tables, worktops, railings, switches, etc.) in a single operation.
Particularly suitable for disinfecting surfaces in medical settings and in sectors such as public buildings, the food and non-food industry or the professional kitchen. Product approved for food contact.
Certifications: Bactericidal: EN1276, EN13697, Bactericide in medical settings: EN13727, Yeasticide: EN1650, EN13697, Yeasticide in medical settings: EN13624, Virucidal against enveloped viruses, Adenovirus and Coronavirus: EN14476.