Order Entry
Belgium
ContactUsLinkComponent
Anti-VIM Mouse Monoclonal Antibody [clone: SPM576]
Anti-VIM Mouse Monoclonal Antibody [clone: SPM576]
N° de catalogue PRSI34-182
Fournisseur:  ProSci Inc.
Anti-VIM Mouse Monoclonal Antibody [clone: SPM576]
N° de catalogue PRSI34-182
Fournisseur:  ProSci Inc.

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    vimentin
  • Symbole de l'antigène:
    VIM
  • Clonalité:
    Monoclonal
  • Clone:
    SPM576
  • Conjugaison:
    Unconjugated
  • Cytométrie en flux:
    Yes
  • Hôte:
    Mouse
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Réactivité:
    Pig,
    Goat,
    Chicken,
    Human,
    Bovine,
    Dog,
    Cat
  • Western blot:
    Yes
  • Format:
    Purified Antibody
  • Formulaire:
    Liquid
  • ID de gène:
    P08670
  • Synonymes antigène:
    VIM
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Température de stockage:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Température de transport:
    4 °C
  • Immunogène:
    Human recombinant protein was used as the immunogen for this VIM antibody.
  • Purification:
    Protein G affinity chromatography
  • Taille:
    100 µg
  • Cdt:
    100 µG

Spécifications

À propos de cet article

This mAb reacts with a 58 kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFPs) such as desmin, keratin, neurofilament, and glial fibrillary acid protein. Anti-vimentin alone is of limited value as a diagnostic tool; however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-vimentin. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain vimentin, e.g. hepatoma and seminoma. Anti-vimentin is also useful as a tissue process control reagent.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Western blot: 0,25-0,5 µg/ml Immunohistochemistry (FFPE): 0,25-0,5 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the VIM antibody for each application should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: SPM576
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Dog, Cat, Pig, Goat, Chicken