Anti-OSBPL10 Rabbit Polyclonal Antibody
PRSI14-846
- Type d'anticorps:Primaire
- Nom de l'antigène:oxysterol binding protein-like 10
- Symbole de l'antigène:OSBPL10
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Western blot:Yes
- Formulaire:Liquid
- ID de gène:114884
- Synonymes antigène:oxysterol-binding protein-related protein 10|OSBP-related protein 10|HGNC:16395|ORP10|oxysterol-binding protein-like protein 10|OSBP9|FLJ20363|OSBPL10|oxysterol binding protein-like 10 [Homo sapiens]
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human OSBPL10 (NP_060254.2)
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: OSBPL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat