Anti-RAB13 Rabbit Polyclonal Antibody
PRSI62-390
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Ras-related protein Rab-13
- Symbole de l'antigène:RAB13
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Réactivité:Human,Mouse
- Western blot:Yes
- Épitope:109-137 aa
- Formulaire:Liquid
- ID de gène:5872
- Modification:None
- Molecular weight:23 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Température de transport:4 °C
- Immunogène:This RAB13 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 109-137 amino acids from the central region of human RAB13.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Cdt:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: RAB13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 109-137
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse