Anti-SELP Mouse Monoclonal Antibody [clone: AK-4] (PE (Phycoerythrin))
Catalog # PRSI76-197
Supplier: ProSci Inc.
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:selectin P (granule membrane protein 140kDa, antigen CD62)
- Symbole de l'antigène:SELP
- Clonalité:Monoclonal
- Clone:AK-4
- Conjugaison:PE (Phycoerythrin)
- Cytométrie en flux:Yes
- Hôte:Mouse
- Isotype:IgG1 kappa
- Réactivité:Human
- Adsorption croisée:No
- Formulaire:liquid
- ID de gène:P16109
- Synonymes antigène:SELP
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Température de stockage:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Température de transport:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cdt:100 Tests
Specifications
About this item
The AK-4 monoclonal antibody specifically reacts with human CD62P (P-Selectin), a 140 kDa type I transmembrane glycoprotein also known as platelet activation-dependent granule membrane protein (PADGEM). Upon activation it is quickly transported to the plasma membrane from the alpha-granules of platelets and WEibel-Palade bodies of endothelial cells. It is essential to the functions of cell adhesion during inflammatory reactions and in the interaction of platelets with monocytes and neutrophils.
Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: AK-4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human