Anti-TAS2R7 Rabbit Polyclonal Antibody
PRSI58-744
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Taste receptor type 2 member 7
- Symbole de l'antigène:TAS2R7
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Réactivité:Human
- Western blot:Yes
- Épitope:208 - 235 aa
- Formulaire:Liquid
- ID de gène:50837
- Modification:None
- Molecular weight:37 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Température de transport:4 °C
- Immunogène:This TAS2R7 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 208-235 amino acids from the C-terminal region of human TAS2R7
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Cdt:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: TAS2R7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 208-235
Host: Rabbit
Isotype: Ig
Reactivity: Human