Order Entry
Belgium
ContactUsLinkComponent
Anti-STX Recombinant Antibody [Clone: SP6]
Anti-STX Recombinant Antibody [Clone: SP6]
N° de catalogue PRSI24-301
Fournisseur:  ProSci Inc.
Anti-STX Recombinant Antibody [Clone: SP6]
N° de catalogue PRSI24-301
Fournisseur:  ProSci Inc.

Spécifications

  • Type d'anticorps:
    Primaire
  • Clonalité:
    Recombinant
  • Clone:
    SP6
  • Conjugaison:
    Unconjugated
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG kappa
  • Réactivité:
    Hamster,
    Pig,
    Rabbit,
    Human,
    Bovine,
    Guinea pig,
    Dog,
    Rat
  • Western blot:
    Yes
  • Adsorption croisée:
    No
  • Formulaire:
    Liquid
  • Synonymes antigène:
    STX
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Température de stockage:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Température de transport:
    Shipped on blue ice
  • Immunogène:
    Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenate
  • Type de conditionnement:
    Vial
  • Cdt:
    0,2 mg

Spécifications

À propos de cet article

Type: Primary
Antigen: STX
Clonality: Recombinant
Clone: SP6
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human;Rat;Hamster;Rabbit;Pig;Dog;Guinea Pig;Bovine