Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PPP
PPP ≥98% (par HPLC)
PPP ≥98% (par HPLC)
PPP ≥98% (par HPLC)
ENZOBMLEI3720001
:
ENZO LIFE SCIENCES
:
PPP ≥98% (par HPLC)
ENZOBMLEI3720001
:
ENZO LIFE SCIENCES
:
Cdt:
1 mg
Seq:
2070
:
:
414.40 g/mol
:
Congélateur
:
477-47-4
:
C₂₂H₂₂O₈
:
MFCD01742647