Order Entry
Belgium
ContactUsLinkComponent
3-(4-Hydroxyphényl)-1-(2,4,6-trihydroxyphényl)propane-1-one ≥98%
3-(4-Hydroxyphényl)-1-(2,4,6-trihydroxyphényl)propane-1-one ≥98%
Catalog # ENZOBMLEI1540200
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
3-(4-Hydroxyphényl)-1-(2,4,6-trihydroxyphényl)propane-1-one ≥98%
Catalog # ENZOBMLEI1540200
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Cdt:
    200 mg
  • Seq:
    2530

Specifications

About this item

 :
 :274.28 g/mol
CAS Number:60-82-2
Formula:C₁₅H₁₄O₅
MDL Number:MFCD00002288