0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Filtres
Filtres à gaz
Gas Filters, Gas Clean kit for 8890 and 8860
Gas Filters, Gas Clean kit for 8890 and 8860
VARI179880
:
VARIAN
Gas Filters, Gas Clean kit for 8890 and 8860
VARI179880
:
VARIAN
Désignation:
Gas clean kit for 8890 and 8860 (includes carrier gas filter, 1/8" connection unit with mounting bracket, and gas clean sensor)
Cdt:
1 KIT