Multitex® Wipes, Loose Wiping Cloth
Catalog # ZVGA10840-00
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
New Product
Specifications
- Emballage:10×40 cloths
- Désignation:Loose wiping cloth
- L×l:300×360 mm
- Cdt:400
Specifications
About this item
Non-woven wipes for disinfection and cleaning.
- 100% PET
- Premium smooth
Tear-resistant, absorbent and low-lint.