PTFE Tubing
Tubing PTFE (Teflon tubing), 0,5 mm, 6,5 mm
  531-1000
PTFE Tubing
  531-1000
PTFE Tubing
PTFE Tubing
Tubing PTFE (Teflon tubing), 0,5 mm, 6,5 mm
10,00 €
1 * 1 ST
  • Tube Ø ext.:
    6,5 mm
  • Tube Ø int.:
    0,5 mm
  • Désignation:
    Tubing PTFE (Teflon tubing)
  • Seq:
    01