Calibration Cuvettes
Calibration cuvettes for UNISTAT® bilirubinometer
N° de catalogue REIC13103410
Fournisseur: REICHERT
Aller à :
Product Details & Documents
Re-usable glass calibration cuvette with special option coatings assayed at a mid-range Total Serum Bilirubin value. Used to perform an automatic calibration on the Reichert UNISTAT® Bilirubinometer.
Approximate Assay Value 20 mg/dL, 342 µmol/L
Spécifications
- Désignation:Calibration cuvettes for UNISTAT® bilirubinometer
- Seq:02