Acide chlorhydrique 0.1 mol/l (0.1 N)
Catalog # FFCHFF2165000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Cdt:5 L
- Seq:5190
Specifications
About this item
:
Density:1,004 g/cm³ (25 °C)
Storage Temperature:Température ambiante
EINECS:231-595-7
ADR:8,III
:36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324