Anti-Gab2 (Tyr452) Phosphate Rabbit Monoclonal Antibody [clone: C33G1]
Type: Primary
Antigen: Gab2 (Tyr452) phosphate
Clonality: Monoclonal
Clone: C33G1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- CSTA3882T
- CSTA3882S
- Symbole de l'antigène:
- Gab2 (Tyr452) phosphate
- Gab2 (Tyr452) phosphate
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Monoclonal
- Monoclonal
- Clone:
- C33G1
- C33G1
- Réactivité:
- Human, Mouse
- Human, Mouse
- Hôte:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Sequence:
- CGRSDSTNSEDNyVPM
- CGRSDSTNSEDNyVPM
- Température de stockage:
- −20 °C
- −20 °C
- Température de transport:
- Ambient
- Ambient
Frequently Bought Together