Anti-Integrin beta4 Rabbit Monoclonal Antibody [clone: D8P6C]
Type: Primary
Antigen: Integrin beta4
Clonality: Monoclonal
Clone: D8P6C
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- CSTA14803T
- CSTA14803S
- Symbole de l'antigène:
- Integrin beta4
- Integrin beta4
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Monoclonal
- Monoclonal
- Clone:
- D8P6C
- D8P6C
- Réactivité:
- Human
- Human
- Hôte:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChimie:
- Yes
- Yes
- Cytométrie en flux:
- Yes
- Yes
- Sequence:
- CDGDSPESRLTVPGLSENVPYKFKV
- CDGDSPESRLTVPGLSENVPYKFKV
- Température de stockage:
- −20 °C
- −20 °C
- Température de transport:
- Cold
- Cold
Frequently Bought Together