MYPT1 Antibody Sampler Kit
N° de catalogue CSTA5143T
Fournisseur: Cell Signaling
Spécifications
- Température de stockage:−20 °C
- Température de transport:Cold
- Cdt:1 KIT
Spécifications
À propos de cet article
The MYPT1 antibody sampler kit is an economical way to examine the total protein levels of MYPT1 as well as MYPT1 phosphorylated at Ser507, Ser668, Thr853, and Thr696. The kit includes enough primary and secondary antibodies to perform two Western blot experiments.
Each antibody in the MYPT1 antibody sampler kit detects endogenous levels of its target protein. Activation state antibodies detect only target proteins phosphorylated at indicated residues.
Livraison: Kit includes: Phospho-MYPT1 (Ser507) Antibody (CSTA3040) 20 µl, Phospho-MYPT1 (Ser668) Antibody (CSTA3048) 20 µl, Phospho-MYPT1 (Thr853) Antibody (CSTA4563) 20 µl, Phospho-MYPT1 (Thr696) Antibody (CSTA5163) 20 µl, MYPT1 (D6C1) Rabbit mAb (CSTA8574) 20 µl, Anti-rabbit IgG 20 µl, HRP-linked Antibody (CSTA7074) 100 µl