Anti-Mic-1/GDF15 Rabbit Monoclonal Antibody [clone: E4D3K]
Catalog # CSTA48481S
Supplier: Cell Signaling
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:Mic-1/GDF15
- Clonalité:Monoclonal
- Clone:E4D3K
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Sequence:Q99988 P2-I308
- Température de stockage:–20 °C
- Température de transport:Ambient
- Cdt:100 µl
Specifications
About this item
Type: Primary
Antigen: Mic-1/GDF15
Clonality: Monoclonal
Clone: E4D3K
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human