Anti-hnRNP P2 Rabbit Monoclonal Antibody [clone: E3O8I]
Catalog # CSTA67840S
Supplier: Cell Signaling
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:hnRNP P2
- Clonalité:Monoclonal
- Clone:E3O8I
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Précipitation immunologique:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CQQSYNPPQGYGQQN
- Température de stockage:–20 °C
- Température de transport:Ambient
- Tested applications:eCLIP
- Cdt:100 µl
Specifications
About this item
Type: Primary
Antigen: hnRNP P2
Clonality: Monoclonal
Clone: E3O8I
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat