Anti-tPP Rabbit Monoclonal Antibody [clone: D61C3]
CSTA5757S
- Type d'anticorps:Primaire
- Symbole de l'antigène:tPP
- Clonalité:Monoclonal
- Clone:D61C3
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Multiple
- Western blot:Yes
- Sequence:CXXXXXtPPXXXX
- Température de stockage:–20 °C
- Température de transport:Cold
- Cdt:100 µl
Type: Primary
Antigen: tPP
Clonality: Monoclonal
Clone: D61C3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species