Anti-NBA1 Rabbit Monoclonal Antibody [clone: D7Y5K]
Catalog # CSTA12711S
Supplier: Cell Signaling
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:NBA1
- Clonalité:Monoclonal
- Clone:D7Y5K
- Hôte:Rabbit
- Précipitation immunologique:Yes
- Isotype:IgG
- Réactivité:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CQSHASYSLLEEEDEA
- Température de stockage:–20 °C
- Température de transport:Ambient
- Cdt:100 µl
Specifications
About this item
Type: Primary
Antigen: NBA1
Clonality: Monoclonal
Clone: D7Y5K
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey