Order Entry
Belgium
ContactUsLinkComponent
 
Anti-DARPP-32 (Thr75) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA2301S
Supplier:  Cell Signaling
Anti-DARPP-32 (Thr75) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA2301S
Supplier:  Cell Signaling

Specifications

  • Type d'anticorps:
    Primaire
  • Symbole de l'antigène:
    DARPP-32 (Thr75) phosphate
  • Clonalité:
    Polyclonal
  • Hôte:
    Rabbit
  • Réactivité:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Température de stockage:
    −20 °C
  • Température de transport:
    Cold
  • Taille:
    100 µl
  • Cdt:
    100 µl

Specifications

About this item

Type: Primary Antibodies
Antigen: DARPP-32 (Thr75) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Mouse,Rat