Acétonitrile
Flacon Verre, 1 L
ROTHHN40.1
Marketsource
: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- :75-05-8
- :H₃CCN
- :MFCD00001878
- :41.05 g/mol
- :13,00071
- :81,6 °C (1013 hPa)
- :–45,7 °C
- :0,782 g/cm³ (20 °C)
- :2 °C (coupe fermée)
- :Température ambiante
- :200-835-2
- :1648
- :3,II
- :01-2119471307-38
- Type de conditionnement:Flacon Verre
- Cdt:1 L
- Cat. No.:ROTHHN40.1
- Certification environnementale:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



