LysoBrite™ Red
LysoBrite™ red, 500 Tests
N° de catalogue AATB22645
Fournisseur: AAT Bioquest
Aller à :
Product Details & Documents
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Lysosome-specific dye
- Long-term use
- Low cytotoxicity
Attention: For research use only.
Spécifications
Spécifications
- Cdt:500 Tests
- Désignation:LysoBrite™ red
- Cat. No.:AATB22645
- Purity:95%