ContactUsLinkComponent
Amplite® Universal Fluorimetric MMP Activity Assay Kit *Green Fluorescence*
Amplite® Universal Fluorimetric MMP Activity Assay Kit *Green Fluorescence*
N° de catalogue AATB13510
Fournisseur:  AAT BIOQUEST
Numéro CAS:  
undefined
Amplite® Universal Fluorimetric MMP Activity Assay Kit *Green Fluorescence*
N° de catalogue AATB13510
Fournisseur:  AAT BIOQUEST
Numéro CAS:  
Spécifications
  • Cdt:
    100 Tests
  • Désignation:
    Amplite™ universal fluorimetric MMP activity assay kit *Green Fluorescence*
Spécifications
À propos de cet article
The matrix metalloproteinases (MMPs) constitute a family of zinc-dependent endopeptidases that function within the extracellular matrix.
  • Bright signal
  • Consistent results
  • In-house quality testing
Attention: For research use only.