Helixyte™ Green Fluorometric dsDNA Quantitation kit *Optimised For Microplate Readers*
N° de catalogue AATB17650
Fournisseur: AAT BIOQUEST
Numéro CAS:
Spécifications
- Cdt:200 Tests
- Désignation:Helixyte™ green fluorimetric dsDNA quantitation kit *optimized for microplate readers*
Spécifications
À propos de cet article
Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and free nucleotides.
- High sensitivity
- Exceptional DNA affinity
- Robust protocol
- Multiple applications
Attention: For research use only.