FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml)
N° de catalogue AATB20005
Fournisseur: AAT Bioquest
Numéro CAS:
Spécifications
- Cdt:20 mL
- Désignation:FluoroQuest™ Anti-fading Mounting Medium with DAPI
Spécifications
À propos de cet article
FluoroQuest™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
- Preserves fluorescence
- Prevents photobleaching
- Contains nuclear counterstain for convenience
Attention: For research use only.