Calcein Red™ sodium salt
1 mg
N° de catalogue AATB21901
Fournisseur: AAT Bioquest
Aller à :
Product Details & Documents
Calcein AM is one of the most popular fluorescent probes used for labeling and monitoring cellular functions of live cells.
- In-house quality control
- long-wavelength emission signal
- Multiple applications
Attention: For research use only.
Spécifications
Spécifications
- Cdt:1 mg