ContactUsLinkComponent
Live or Dead™ Fixable Dead Cell Staining Kit *Green Fluorescence with 405 nm Excitation*
Live or Dead™ Fixable Dead Cell Staining Kit *Green Fluorescence with 405 nm Excitation*
Catalog # AATB22501
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Live or Dead™ Fixable Dead Cell Staining Kit *Green Fluorescence with 405 nm Excitation*
Catalog # AATB22501
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Cdt:
    200 Tests
  • Désignation:
    Live or Dead™ fixable dead cell staining kit *green fluorescence with 405 nm excitation*
  • Tests par set:
    200 Tests
Specifications
About this item
Our Live or Dead™ Fixable Dead Cell Staining Kits are a set of tools for labeling cells for fluorescence microscopic investigations of cellular functions.
  • Contains all necessary components
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Large stokes shift
Caution: For research use only.