Disinfectant, Anioxyde 1000 LD
N° de catalogue DMDA2824299EV
Fournisseur: Laboratoires Anios
Spécifications
- Capacité:5 L
- Désignation:Anioxyde 1000 LD with integrated activator
- Emballage:Canister
- Cdt:4
Spécifications
À propos de cet article
High-level disinfectant for thermosensitive instruments and endoscopy equipment.
- Shelf life before use: 24 months
- High level disinfectant from PHERA® system, active against spores of bacteria
- Extemporaneous production of peracetic acid
- Alternative to glutaraldehyde
- Aldehyde free: no fixation of proteins
- Possible control of bath efficacy with test-strips
- Total effectiveness in 5 minutes
Attention: Use biocides safely. Always read the label and product information before use.
For professional use only.
For professional use only.