Monoject™ Blunt Cannulas with Aluminium Hub
Fournisseur: CARDINAL HEALTH
New Product
Certificats
À propos de cet article
Monoject™ blunt cannula is an individually packaged blunt needle. Features include sterile, disposable, rigid pack packaging, stainless steel cannula and aluminium Luer-lock hub.
- Stainless steel, non-lubricated cannula
- Rounded blunt end
- Aluminum luer-lock hub
- Rigid pack packaging with heat stake, cap-to-sheath seal
- Not manufactured with DEHP or natural rubber latex
- Sterile, single-use and individually packaged
Frequently Bought Together