0
0,00 €
Order Entry
Connexion
/
S'inscrire
Products
Services
Digital Solutions
Applications
Support
About
Saisie de commande
Statut de la commande
Votre compte
Panier
Belgium
Saisie de commande
Statut de la commande
Votre compte
Panier
ContactUsLinkComponent
Accueil
Enzymes
Recombinant Tobacco Etch Virus (from E. coli)
Recombinant Tobacco Etch Virus (from E. coli)
Recombinant Tobacco Etch Virus (from E. coli)
Catalog # ABNOP7329.1000IU
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Tobacco Etch Virus (from E. coli)
Catalog # ABNOP7329.1000IU
Supplier:
Abnova
CAS Number:
Specifications
Cdt:
1.000 AU
Type d'enzyme:
Recombinant
Source:
E. coli
Nom de l'enzyme:
Tobacco Etch Virus
Specifications
About this item
Product Details
Product Details