ContactUsLinkComponent
Beakers with Spout (316L Stainless Steel)
  SAMPA866-1000
 : A866-1000
undefined
Beakers with Spout (316L Stainless Steel)
  SAMPA866-1000
 : A866-1000
Select an Option
1000 ml
Select an Option
Bécher avec bec verseur
Select an Option
04
Select an Option
1
These beakers with spout are excellent for handling and dispensing a variety of liquids.
  • Made from high grade 316L stainless steel
  • Excellent corrosion resistance
  • Wide range of sizes available
These stainless steel beakers with spout are made from top quality 316L stainless steel which provides excellent corrosion resistance and provides an ideal way to store, handle and dispense a wide variety of liquids across many industries.
Frequently Bought Together