μPAC™ ESI-MS Accessories
N° de catalogue HYPEEMI-EASYSPRAYB
Fournisseur: Thermo Fisher Scientific
Spécifications
- Pour:EASY-Spray™ source
- Désignation:μPAC™ compatible EASY-spray emitter
- Cdt:1
Spécifications
À propos de cet article
ESI-MS accessories are used to facilitate reproducible, low dispersion connections between μPAC HPLC columns and low-flow mass spectrometer ion sources.
The μPAC EASY-spray emitter is equipped with a stainless steel 50 μm through-bore union. Achieve high resolution separations in low-flow applications when using this emitter to connect a μPAC HPLC column and a transfer line with a EASY-Spray™ source.
The μPAC EASY-spray emitter is designed for easy connection to an EASY-Spray MS source. It contains a pre-assembled EASY-Spray emitter with a transfer line and grounding union.