CSLAB Interchangeable Syringes
Product Details & Documents
These Cadence Science® CSLAB interchangeable syringes are manufactured to ensure the perfect mating of barrels and plungers. Any interchangeable syringe of a particular size will fit any interchangeable barrel of that same size.
- Precision shrinking ensures improved fit
- Coloured plunger ends are fused around bottom beveled rim of plunger to facilitate dosage measurements
- Made from borosilicate glass providing protection against thermal shock
- Corrosion resistant
- Graduations permanently fused into glass barrel and reinforced flanges prevent syringe rolling
- Chrome-plated rings give the user a positive grip, permitting easy and accurate delivery
CS Lab matched numbered syringes feature frosted individually lapped barrels and plungers which can withstand higher temperatures. They are individually boxed and have matching serial numbers on mating barrels and plungers. CSLAb specialty matched numbered syringe features an 0,088" effluent luer lock tip which ensures that a greater fluid rate can be achieved during the injection. This specially designed syringe has ground glass barrels featuring a reduced barrel ID. This reduction produces higher pressures during injection.
CSLab matched numbered tuberculin syringes are fabricated of chemically resistant borosilicate glass with easy-to-read accurate permanent calibration lines. They are widely used in lab and research procedures for the administration of precise low-volume doses. CSLab pressure control are designed with large comfortable thumb and finger rings. They are particularly useful for injecting viscous fluids or in any non-medical application that requires one-hand filling.
Caution: Not for medical/human use.
Specifications
- Désignation:Glass syringe with glass tip, ¹/₄ ml in ¹/₁₀₀ ml volume
- Matière:Verre borosilicaté
- Seq:05