IQ/OQ Documentation for the OC Ampoule Opener and Closer
Catalog # ADLM6035000-IQOQ
Supplier: ADELPHI MANUFACTURING
Specifications
- Désignation:IQ/OQ documentation for the OC ampoule opener and closer
- Cdt:1
Specifications
About this item
IQ/OQ documentation for the Adelphi Manufacturing OC ampoule opener and closer.
- Installation Qualification and Operational Qualification documentation
- The OC machine itself enables opening and sealing of closed-type ampoules
- Although specially designed to handle closed-type ampoules, the OC will also handle open-type ampoules after washing
Please note that this listing only refers to the IQ/OQ Documentation, which accompanies the Adelphi Manufacturing OC ampoule opener and closer.
Installation Qualification: Performed to ensure that all key aspects of the process equipment and ancillary system installation adhere to the manufacturer's approved specification and that the recommendations of the supplier of the equipment (usually according to the installation manual) are suitably considered.
Operational Qualification: Checks and documents that each individual function of the item operates as expected. For example, running at the required speed or heating up a liquid to the required temperature.