TAAG E41 Universal Pathogens
TAAG E41 universal pathogens, V-FE04-1, 1 jar - 500 g, 1
N° de catalogue TAAGTGG0003
Fournisseur: KREGLINGER EUROPE
Aller à :
Product Details & Documents
Dehydrated enrichment media for growing Salmonella spp., Staphylococcus aureus, Escherichia coli and Listeria monocytogenes in food samples. It contains selected growth factors and nutritional requirements optimized to promote pathogens development.
Spécifications
Spécifications
- Modèle:V-FE04-1
- Cdt:1
- Désignation:TAAG E41 universal pathogens
- Contenu:1 jar - 500 g