VWR® Safety Shower Testing Unit
VWR® Safety Shower Testing Unit
Safety shower testing unit
N° de catalogue 139-0806
Fournisseur:   VWR Collection
VWR® Safety Shower Testing Unit
N° de catalogue 139-0806
Fournisseur:  VWR Collection
VWR® Safety Shower Testing Unit
VWR® Safety Shower Testing Unit
Safety shower testing unit
Prix
2 650,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Laboratory safety showers should be tested once a month to check the functionality and the required water flow rate of 60 L/min. for emergency instances. This mobile test unit provides a very convenient and easy solution to test both body safety showers and handheld eye showers.
  • Transparent PVC water reservoir on castors
  • 110 litre capacity
  • Built-in suction pump (230 V) and IP 45 protected power switch
  • Drain hose (2 m) to empty the water reservoir
  • Separate mounts for drain hose and power cord
Placed centrally under the body safety shower, the large funnel can be extended to a height of 2,30 metres, while the smaller funnel is designed to collect the water from handheld eye showers (minimum flow rate: 6 L/min). The collected water amount can be read on the water reservoir scale from 0 to 110 litres. The built-in electrical suction pump will empty the reservoir in a very short time. According to EN 15154-1,2:2006 the water flow rate must be at least 60 L/min. for plumbed-in body showers and 6 L/min. for plumbed-in eye wash units.
Spécifications
  • Désignation:
    Safety shower testing unit