pH Sensor Pure, Memosens CPL57E
Product Details & Documents
Potentiometric pH glass sensor for laboratory measurement and random sampling in pure water. Connectable via inductive, digital connection with Memosens technology.
- pH sensor for boiler feed water and water with low conductivity
- High accuracy and long-time stability in pure water thanks to additional KCL salt storage
The Memosens 2.0 pH sensor CPL57E is specially designed for pure water applications in all industries. This glass sensor offers long-term stability, thanks to the additional salt storage, and ensures highly accurate measured values in pure water.
Thanks to Memosens technology, the sensors provide reliable, highly accurate measurements with a fast response time.
The inductive Memosens coupling between the corresponding Memosens sensor and the mobile CML18 handheld device is insensitive to moisture and corrosion.
The Memosens technology is based on a true plug-and-play principle. Just connect the sensor and you are ready to start your measurements. Measured values, calibration data and history are stored directly in the sensor. This means status values can be read out and a sensor check is also possible. An excellent feature to boost your GLP.
All sensors are supplied pre-calibrated. Thus, they are directly ready for use for your random sample measurement when the corresponding sensor is connected to the associated mobile handheld device Liquiline Mobile CML18.
Certifications: EC_00828_03.22; EU_01000_01.22; UK_00482_01.24. Nonhazardous area appoval, CE + UKCA marking.
Documents
Spécifications
- Type:Potentiometric pH glass sensor
- Désignation:Memosens CPL57E pH sensor CPL57E Pure, 120 mm; Teflon ring junction, salt store, saturated KCl, Ag/AgCl ref. system
- pH:2 - 12 pH, –5…100 °C
- Plage de pH:1 - 12
- Cat. No.:662-2460
- Type de sonde:Basic version, zero point pH 7,0, temperature sensor NTC 30k
- Reference system:Teflon ring junction, salt store, saturated KCl, Ag/AgCl
- Connexion:Memosens
- Gamme:2 - 12 pH, –5…100 °C (application range)