VWR®, Septa ND12
548-0185A
- Septum:Universal HT injection port, transparent
- Épaisseur:3,0 mm
- Cdt:25
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
- Fits for a broad range of caps
- Easy exchange
- Different hardness and septa thickness available
Septa choice depends on the application.